Polyethylene glycol monostearate 400
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Polyoxyethylene stearate, Polyoxyl stearate
Polyethylene Glycol 400 Monostearate, commonly referred to as PEG 400 MONOSTEARATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
HO(CH₂CH₂O)nOCC₁₇H₃₅ Storage Temperature: Ambient |
MDL Number:
MFCD00148007 CAS Number: 9004-99-3 |
Specification Test Results
Saponification Value | 83 - 92 |
Hydroxyl Value | 80 - 100 |
pH (5% Solution) | 5.0 - 7.0 |
Identification | To Pass Test |
Acid Value | max. 2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...