Castor oil, hydrogenated, ethoxylated 40, hydrogenated

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethoxylated hydrogenated castor oil, Kolliphor® RH, Macrogolglycerol hydroxystearate, PEG castor oil, Cremophor® RH, Polyethylene glycol castor oil hydrogenated, Polyoxyl hydrogenated castor oil

P2132-500GM P2132-18KG P2132-2.5KG
700015-350EA 132.39 USD
700015-350 700015-344 700015-346
Castor oil, hydrogenated, ethoxylated 40, hydrogenated
Castor oil, hydrogenated, ethoxylated

Polyethylene Glycol 40 Castor Oil, Hydrogenated is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD01771439
CAS Number: 61788-85-0

Order Now

Specification Test Results

Appearance @ 25° C Yellow Liquid To
Saponification Number 60 - 87
Hydroxyl Value 59 - 68
Color (Gardner) max. 6
Moisture (KF) max. 1.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR