Polyethylene glycol 1,450 NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: PEG 1,450, Ethylene glycol polymer 1450, PEG 1450

P1492-2.5KG P1492-500GM P1492-12KGBL P1492-45KGBL
95034-986EA 296.05 USD
95034-986 95034-984 75811-970 75811-972
Polyethylene glycol 1,450 NF
Polyethylene glycol 1,450

Polyethylene Glycol 1450, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: H(OCH₂CH₂)nOH
MW: 1450 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

Average Molecular Weight 1305 - 1595
Completeness and Color of Solution To Pass Test
Actual Viscosity @ 98.9 C+/-0.3 C (210 F) 25 - 32 cSt
pH of a 5% Solution @ 25°C 4.5 - 7.5
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR