Pimenta berry oil

Supplier: SPECTRUM CHEMICAL MFG CORP

P1131-125ML P1131-22.5KG P1131-25ML P1131-500MLG
700010-762EA 549.5 USD
700010-762 700010-764 700010-766 700010-768
Pimenta berry oil
Pimenta berry oil

Pimenta Berry Oil is a flavor and fragrance ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00240716
CAS Number: 8006-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR