Phloxin B

Supplier: SPECTRUM CHEMICAL MFG CORP

PH170-100GM PH170-10GM PH170-25GM
700010-140EA 681.05 USD
700010-140 700010-142 700010-144
Phloxin B
Phloxin B

Phloxine B, also known simply as phloxine, is used in the hematoxylin-phloxine-saffron stain.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₂Br₄Cl₄Na₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00070627
CAS Number: 18472-87-2

Order Now

Specification Test Results

Absorption Max 548 (510)NM
Visual Transition Intervals pH 2.1 (Clrls.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR