Peru balsam (Balsamum peruvianum)

Supplier: SPECTRUM CHEMICAL MFG CORP

PE120-125GM PE120-2.5KG PE120-500GM
700010-790EA 151.3 USD
700010-790 700010-792 700010-794 700010-796
Peru balsam (Balsamum peruvianum)
Peru balsam (Balsamum peruvianum)

Peruvian Balsam is also known as Balsam of Peru and is derived from the tree grown in El Salvador which is known as Myroxylon. The balsam comes from the trunk of the tree which is similar to substances like vanilla, clove and cinnamon.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 1.161
Storage Temperature: Ambient
MDL Number: MFCD00147893
CAS Number: 8007-00-9

Order Now

Specification Test Results

Analysis By GC To Pass Test
Refractive Index @ 20°C 1.5800 - 1.6000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR