Peppermint oil NF

Supplier: SPECTRUM CHEMICAL MFG CORP

PE105-500MLG PE105-10KGBL PE105-100ML PE105-25ML
700010-412EA 594.64 USD
700010-412 89078-918 95034-856 95034-858
Peppermint oil NF
Peppermint oil

Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Storage Temperature: Ambient
MDL Number: MFCD00147870
CAS Number: 8006-90-4

Order Now

Specification Test Results

Esters(As Menthyl Acetate) 5.0 % min.
Total Menthol(Free and As Esters) 50.0 % min.
Specific Gravity @ 25°C 0.896 - 0.908
Angular Rotation -18° to -32°
Refractive Index @ 20°C 1.459 - 1.465


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR