Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane, Antioxidant 1010, 2,2-Bis(((3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) is used as primary antioxidant for stabilizing polymers, particularly polyethylene and polypropylene.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇₃H₁₀₈O₁₂ Storage Temperature: Ambient |
MDL Number:
MFCD00059345 CAS Number: 6683-19-8 |
Specification Test Results
Assay | 98% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...