Peanut oil NF
Supplier: Spectrum Chemical Mfg. Corp.
Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
MDL Number:
MFCD00131805 CAS Number: 8002-03-7 |
Specification Test Results
Fatty Acid Composition | To Pass Test |
Acid Value | max.0.2 |
Alkaline Impurities | To Pass Test |
Heavy Metals | max. 10 ppm |
Peroxide Value | max. 5.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...