Alphazurine A

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Fenazo bluexg, Patent Blue A, Carmine Blue AF, Alphazurine A, Erio Glaucine X, Acid Blue 7

PA140-100GM PA140-25GM
700010-136EA 464 USD
700010-136 700010-138
Alphazurine A
Alphazurine A

Patent Blue A is a dye for wool, silk, polyamide, soaps, and cosmetics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₇H₃₅N₂NaO₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00044440
CAS Number: 3486-30-4

Order Now

Specification Test Results

Absorption Max 637 (409) NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR