4-Aminophenol ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
p-Aminophenol, UrsolP Rodinal 4-Amino-1-hydroxybenzene p-Hyddroxyaniline 4-Aminophenol 4-Hydroxyaniline, 4-Hydroxyaniline
A2305-100GM
A2305-500GM
10841-924EA
69.35
USD
10841-924
10841-926
4-Aminophenol ≥98%
4-Aminophenol
p-Aminophenol, also known as 4-Aminophenol, is a building block compound and can be used as a film developer.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
H₂NC₆H₄OH MW: 109.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007869 CAS Number: 123-30-8 UN: 2512 ADR: 6.1,III |
Specification Test Results
Assay | 98 % min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...