o-(2-Aminopropyl)-o'-(2-methoxyethyl)propylene glycol, M.W. 500 500

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Jeffamine® M-600, Polypropylene glycol mono-2-aminoethyl mono-2-methoxyethyl ether

A2243-100ML A2243-500MLGL
10841-878EA 108.55 USD
10841-878 10841-880
o-(2-Aminopropyl)-o'-(2-methoxyethyl)propylene glycol, M.W. 500 500
o-(2-Aminopropyl)-o'-(2-methoxyethyl)propylene glycol

O-(2-Aminopropyl)-O'-(2-methoxyethyl)propylene Glycol 500 is a polyether that is a nine-membered polypropylene glycol terminated by an amino group at one end and a methoxyethyl termination at the other. It is a common crystallization reagent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.983
Storage Temperature: Ambient
MDL Number: MFCD00147774
CAS Number: 77110-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR