n-Tetradecane ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Tetradecane

T2007-500ML T2007-100ML T2007-2.5LTGL
10860-030EA 388.6 USD
10860-030 10860-032 10860-304
n-Tetradecane ≥99%
n-Tetradecane

GHS hazard : Health hazard;Exclamation mark

Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Density: 0.763
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR