Nicotinamide ≥98.0%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
Niacinamide is also referred to as nicotinic acid amide. A water soluble vitamin that is part of the vitamin B group, it does not have cholesterol reducing effects or cause flushing. Used in medicine to treat skin conditions as it works as an anti inflammatory, it is also employed in cancer treatment by enhancing tumor blood flow as well as being used to prevent apoptosis. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use and typically are not suitable for consumption.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₆N₂O MW: 122.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006395 CAS Number: 98-92-0 |
Specification Test Results
Assay | 98.0-101.0 % |
Melting Range | 128 - 131 °C |
Heavy Metals (As Pb) | max. 0.003 % |
Loss On Drying | max. 0.5 % |
Readily Carbonizable Substances | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...