Sodium decanoate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

D2852-25GM D2852-100GM
700005-114EA 72.5 USD
700005-114 700005-112
Sodium decanoate ≥99%
Sodium decanoate

Storage temperature - Refrigerate


Precaución: Storage temperature - Refrigerate

Formula: C₁₀H₁₉NaO₂
MW: 194.2464 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00066453
CAS Number: 1002-62-6

Order Now

Specification Test Results

Assay Approx. 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR