N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Butanol, 4-(butylnitrosoamino)-, N-Butyl-N-nitroso-4-aminobutanol, N-Butyl-N-butan-4-ol-nitrosamine, BBN

B2500-1GM B2500-5GM B2500-25GM
10860-554EA 171 USD
10860-554 10860-556 10860-558
N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%
N-Butyl-N-(4-hydroxybutyl)nitrosamine

Storage temperature - LIGHT SENSITIVE, AIR SENSITIVE, MOISTURE SENSITIVE, HEAT SENSITIVE: Keep tightly closed in light-resistant containers.


GHS hazard : Exclamation mark;Health hazard


Precaución: Storage temperature - LIGHT SENSITIVE, AIR SENSITIVE, MOISTURE SENSITIVE, HEAT SENSITIVE: Keep tightly closed in light-resistant containers.

Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059008
CAS Number: 3817-11-6

Order Now

Specification Test Results

Purity (GC) 90.0 % min.
Specific Gravity (20/20) 1.0060 - 1.0140
Refractive Index (N20/D) 1.4700 - 1.4740


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR