N-Acetyl-L-tyrosine
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH
A2017-25GM
A2017-25KGBL
A2017-5KGBL
A2017-5GM
700007-802EA
347.3
USD
700007-802
700007-804
700007-808
700007-806
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine, also known as 4-hydroxyphenylalanine, is used by cells to synthesize proteins.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₁H₁₃NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00037190 CAS Number: 537-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...