Nɑ-Acetyl-L-glutamine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N-Acetyl-L-glutamine, N-Ac-Gln-OH, Ac-L-Gln-OH, Ac-Gln-OH

A2393-100KGB A2393-100GM
700007-722EA 28172.9 USD
700007-722 700007-724 700007-720
Nɑ-Acetyl-L-glutamine
Nalpha-Acetyl-L-glutamine

N-Acetyl-L-glutamine is used as a skin conditioning agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₁₂N₂O₄
MW: 188.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038159
CAS Number: 2490-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR