N,N-Dimethyl-p-toluidine ≥98.5%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

D2335-100GM D2335-25GM
10841-914EA 150.4 USD
10841-914 10841-916
N,N-Dimethyl-p-toluidine ≥98.5%
N,N-Dimethyl-p-toluidine

N,N-Dimethyl-p-toluidine is used to make acrylic resins and denture materials.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₃N
Density: 0.937
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98.5 % min.
Moisture max.0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR