Diisopropylcarbodiimide
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC
D3183-100GM
D3183-25GM
10841-898EA
388.76
USD
10841-898
10841-900
Diisopropylcarbodiimide
Diisopropylcarbodiimide
N,N'-Diisopropylcarbodiimide, a carbodiimide, is commonly used in peptide synthesis.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇H₁₄N₂ MW: 126.2 g/mol Density: 0.806 Storage Temperature: Ambient |
MDL Number:
MFCD00065689 CAS Number: 693-13-0 UN: 3384 ADR: 6.1 (3),I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...