N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N,N,N,N-Tetrakis(2-hydroxyethyl)ethylenediamine

T1278-100GM T1278-25GM T1278-500GM
700007-056EA 109.85 USD
700007-056 700007-058 700007-060
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine is a cosmetic buffering and chelating agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₄N₂O₄
MW: 236.3086 g/mol
Density: 1.10
Storage Temperature: Ambient
MDL Number: MFCD00002854
CAS Number: 140-07-8

Order Now

Specification Test Results

Appearance Clear Colorless
Assay (Total Base) 98.5% min.
pH (25% Solution) 10 min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR