Dimethyl sulphone ≥99%, crystals

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methyl sulphone, Dimethylsulphone

M2125-500GM M2125-100GM M2125-50KGBL
700002-508EA 3216.04 USD
700002-508 700002-510 700002-506 700002-512
Dimethyl sulphone ≥99%, crystals
Dimethyl sulphone

Methyl Sulfone, Crystal is also known as sulfonylbismethane or methylsulfonylmethane. It is primarily used as a solvent for organic and inorganic substances because of its polarity and thermal stability.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂H₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD00007566
CAS Number: 67-71-0

Order Now

Specification Test Results

Assay 99 %
Melting Point 107°C min.
Loss On Drying max. 1 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR