α-D-(+)-Melibiose, hydrate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 6-O-alpha-D-Galactopyranosyl-D-glucose

M1142-100GM M1142-25GM M1142-5GM
700009-666EA 2808.4 USD
700009-666 700009-668 700009-670
α-D-(+)-Melibiose, hydrate ≥99%
alpha-D-(+)-Melibiose

Melibiose, Hydrate, a reducing disaccharide, is known to form by an alpha-1,6 linkage between glucose and galactose.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00198188
CAS Number: 585-99-9

Order Now

Specification Test Results

Assay 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR