Mannan

Supplier: SPECTRUM CHEMICAL MFG CORP

M3007-1GM M3007-5GM
700012-940EA 863.68 USD
700012-940 700012-944 700012-942
Mannan
Mannan

Mannan, a plant polysaccharide, is a linear polymer of the sugar mannose.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Refrigerator
MDL Number: MFCD00131574
CAS Number: 9036-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR