Manganese glycerophosphate ≥98.0%, Powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

MA163-12KG MA163-500GM MA163-2.5KG MA163-50KGBL
700014-876EA 4816.5 USD
700014-876 95034-316 95034-314 700014-878 95034-312
Manganese glycerophosphate ≥98.0%, Powder FCC
Manganese glycerophosphate

Manganese Glycerophosphate, Powder, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 1320-46-3

Order Now

Specification Test Results

Assay (C₃H₇MnO₆P) 98.0-100.5 %
Loss On Drying max.12.0 %
Lead (Pb) max. 4 mg/kg
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR