Magnesium stearate FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
M1184-2.5CS4
M1184-2.5KG
M1184-500CS6
M1184-500GM
M1184-45KGBL
95034-248CS
1802.95
USD
95034-248
95034-250
95034-252
95034-254
75811-600
Magnesium stearate FCC
Magnesium stearate
Formula:
C₃₆H₇₀MgO₄ |
MDL Number:
MFCD00036391 CAS Number: 557-04-0 |
Specification Test Results
Assay (MgO) | 6.8 - 8.3 % |
Lead (Pb) | max.5 mg/kg |
Loss On Drying | max. 4.0 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...