Magnesium aluminum silicate NF Type IA
Supplier: Spectrum Chemical Mfg. Corp.
Magnesium Aluminum Silicate, Type IA, NF is used in the pharmaceutical formulation process as an absorbent, anticaking agent, opacifying agent, slip modifier, and an aqueous viscosity increasing agent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
CAS Number:
1327-43-1 |
Specification Test Results
Viscosity @ 33 +/- 3 °C | 225 - 600 cP |
pH of a 5 in 100 Suspension | 9.0 - 10.0 |
Ratio of Content Aluminum / Magnesium | 0.5 - 1.2 |
Acid Demand | max. 4.0 |
Loss On Drying | max. 8.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...