L-Tyrosine methyl ester hydrochloride ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
L-Methyltyrosinate hydrochloride, H-Tyr-OMe.HCl, H-Tyr-OMe·HCl
T3045-25GM
T3045-100GM
700007-488EA
106.55
USD
700007-488
700007-486
L-Tyrosine methyl ester hydrochloride ≥98%
L-Tyrosine methyl ester hydrochloride
L-Tyrosine Methyl Ester Hydrochloride is used as an amino acid synthetic reagent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₃NO₃ MW: 231.68 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00012607 CAS Number: 3417-91-2 |
Specification Test Results
Assay | 98 % |
Melting Point | 185 - 195 °C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...