Locust bean gum FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Locust (carob) bean gum
Locust (Carob) Bean Gum, FCC is a gelling and thickening agent used in certain foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00131257 CAS Number: 9000-40-2 |
Specification Test Results
Galactomannans | 75.0 % |
Acid-Insoluble Matter | max.4.0 % |
Arsenic (As) | max. 3 mg/kg |
Ash (Total) | max. 1.2 % |
Lead (Pb) | max. 5 mg/kg |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...