L-Aspartic acid monosodium salt monohydrate ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate

A1375-25GM A1375-50KGBL A1375-1KG
700013-468EA 182.32 USD
700013-468 700013-472 700013-474 700013-470
L-Aspartic acid monosodium salt monohydrate ≥98.0%
L-Aspartic acid monosodium salt monohydrate

L-Aspartic Acid Sodium Salt, Monohydrate is used as a principal neurotransmitter for fast synaptic excitation.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₆NNaO₄·H₂O
MW: 173.11 g/mol (155.10 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00152960
CAS Number: 323194-76-9

Order Now

Specification Test Results

Assay 98.0 - 101.0 %
Specific Rotation [A]D +19.0 to +21.0
Transmittance 95.0 %
Chloride (Cl) max. 0.04 %
Ammonium (NH₄) max. 0.04 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR