Lactulose ≥99%, Powder

Supplier: Spectrum Chemical Mfg. Corp.

L1247-25GM L1247-5GM L1247-5KGBL L1247-100GM
700009-660EA 225.95 USD
700009-660 700009-662 700009-664 700009-658
Lactulose ≥99%, Powder
Lactulose

Lactulose, Powder, a synthetic sugar, is used to treat hepatic encephalopathy and chronic constipation. It is classified as an osmotic laxative. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₂H₂₂O₁₁
Storage Temperature: Ambient
MDL Number: MFCD00151469
CAS Number: 4618-18-2

Order Now

Specification Test Results

Assay 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR