Lactobionic acid, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid

L1017-5GM L1017-10KGBL L1017-100GM
700008-524EA 107.15 USD
700008-524 700008-522 700008-520
Lactobionic acid, Reagent Grade
Lactobionic acid

Lactobionic Acid, Reagent, also known as Galactosylgluconic acid, is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₂H₂₂O₁₂
MW: 358.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078147
CAS Number: 96-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR