Karaya gum FCC
Supplier: Spectrum Chemical Mfg. Corp.
Karaya Gum, FCC, a vegetable gum, is used as a thickener and emulsifier in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00131251 CAS Number: 9000-36-6 |
Specification Test Results
Ash (Acid-Insoluble) | max.1.0 % |
Insoluble Matter | max. 3.0 % |
Lead (Pb) | max. 2 mg/kg |
Loss On Drying | max. 20.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...