Isobornyl acetate ≥90.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

I2076-100ML I2076-500ML
700004-520EA 91.41 USD
700004-520 700004-522
Isobornyl acetate ≥90.0%
Isobornyl acetate

Isobornyl Acetate is used in perfuming soaps, air fresheners and in making camphor. It is also used as a flavoring ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₂₀O₂
Density: 0.983
Storage Temperature: Ambient
MDL Number: MFCD00135943
CAS Number: 125-12-2

Order Now

Specification Test Results

Assay 90.0 % min.
Specific Rotation, [A]D20 -2.0° to +2.0°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR