Isoamyl salicylate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isopentyl 2-Hydroxybenzoate, Isopentyl Salicylate, Salicylic Acid Isoamyl Ester, Isopentyl 2-hydroxybenzoate

I1302-25KGBL I1302-500GM I1302-2.5KG
700006-414EA 1743.45 USD
700006-414 700006-416 700006-412
Isoamyl salicylate
Isoamyl salicylate

Isoamyl Salicylate is a flavor and fragrance agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00020037
CAS Number: 87-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR