Imidazolidinyl urea NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Imidurea, 1,1'-Methylenebis(3-(3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)urea), 1,1'-Methylenebis[3-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]urea], N,N'-Methylenebis[N'-(3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl)urea]

I1007-12KG I1007-125GM I1007-2.5KG I1007-50KGBL I1007-500GM
700008-640EA 2855.3 USD
700008-640 95033-682 95033-684 700008-642 95033-686
Imidazolidinyl urea NF
Imidazolidinyl urea

IMIDUREA, NF is also referred to as imidazolidinyl urea and is used in cosmetics as an antimicrobial preservative that is chemically related to diazolidinyl urea. It is used as a formaldehyde releaser and in its commercial form is a mixture of various formaldehyde addition products that can include polymers. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₁H₁₆N₈O₈
MW: 388.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221482
CAS Number: 39236-46-9

Order Now

Specification Test Results

Assay (As Nitrogen) 26.0 - 28.0 %
Color and Clarity of Solution To Pass Test
pH of a 1% Solution 6.0 - 7.5
Loss On Drying max. 3.0 %
Residue On Ignition max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR