Hydroxypropyl cellulose, 75 - 150 cP (5 % in H₂O) NF

Supplier: SPECTRUM CHEMICAL MFG CORP

HY117-500GM HY117-125GM HY117-12KG HY117-2.5KG
95033-600EA 400.2 USD
95033-600 95033-596 700013-022 95033-598
Hydroxypropyl cellulose, 75 - 150 cP (5 % in H₂O) NF
Hydroxypropyl cellulose

Hydroxypropyl Cellulose, 75-150 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00132688
CAS Number: 9004-64-2

Order Now

Specification Test Results

Assay (Hydroxypropoxy Groups) 53.4 - 80.5 %
Apparent Viscosity of a 5% Aqueous Soln. @25C As Reported
pH of a 1% Solution 5.0 - 8.0
Loss On Drying max. 5.0 %
Silica max. 0.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR