Hydroxyethyl cellulose, 100 cP NF

Supplier: Spectrum Chemical Mfg. Corp.

HY112-45KGBL HY112-12KG HY112-100GM HY112-2.5KG HY112-500GM
700013-020EA 8133.7 USD
700013-020 700013-018 95033-566 95033-568 95033-570
Hydroxyethyl cellulose, 100 cP NF
Hydroxyethyl cellulose

Hydroxyethyl Cellulose, 100 cP, NF. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now

Specification Test Results

Viscosity of a 5% Aqueous Solution 75 - 150 cPs
pH of a 1 % Solution @ 25°C 6.0 - 8.5
Loss On Drying max. 10.0 %
Heavy Metals max. 20 Ug/G
Lead (Pb) max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR