Histamine dihydrochloride ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.

HI115-5GM HI115-25GM HI115-100GM HI115-1KGBL
700008-638EA 85.11 USD
700008-638 700008-636 700008-632 700008-634
Histamine dihydrochloride ≥98%, Reagent Grade
Histamine dihydrochloride

Histamine Dihydrochloride, Reagent is used to give relief of a temporary nature to aches, muscles and joint pain, sprains and other arthritis related ailments. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.


GHS hazard : Exclamation mark;Health hazard

Formula: C₅H₁₁Cl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD00012703
CAS Number: 56-92-8

Order Now

Specification Test Results

Assay 98 % min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR