Hectorite

Supplier: SPECTRUM CHEMICAL MFG CORP

H1406-2KG H1406-500GM
700014-500EA 625.21 USD
700014-500 700014-502
Hectorite
Hectorite

Hectorite, a rare white clay mineral, is found in the brown clay ghassoul which is used in making cosmetics and as a mineral source for refined lithium metal.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: Na₀.₆₇(Mg,Li)₆Si₈O₂₀(OH,F)₄
Storage Temperature: Ambient
CAS Number: 12173-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR