Glyceryl monooleate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Oleoyl-Rac-Glycerol, Glycerol mono-oleate, Mono-Olein

G1017-2.5LTG G1017-200LTB G1017-20LT G1017-500MLG
700014-324EA 410.25 USD
700014-324 700014-326 700014-328 700014-330
Glyceryl monooleate
Glyceryl monooleate

Glyceryl Monooleate is a clear or light yellow oil that is used as an antifoam in juice processing. It has been used as an emulsifier, a moisturizer, and a flavoring agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₄₀O₄
MW: 356.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042735
CAS Number: 25496-72-4

Order Now

Specification Test Results

Saponification Value 166 - 174
Acid Value max. 5
Specific Gravity @ 25°C 0.90-1.00


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR