Tribehenin

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Glycerol Tridocosanoate, Glyceryl tribehenate, Glyceryl behenate, 1,2,3-Tridocosanoyl Glycerol

G1109-5GM G1109-1GM
700014-360EA 909 USD
700014-360 700014-358
Tribehenin
Tribehenin

Glyceryl Behenate, also known as Glycerol docosanoate, is a fat that is used as a viscosity-increasing agent in emulsions in foods & cosmetics, and as a tablet and a lubricant & lipidic coating excipient in oral pharmaceutical formulations.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆₉H₁₃₄O₆
MW: 1059.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056274
CAS Number: 18641-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR