D-Glucono-1,5-lactone ≥99.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
Gluconolactone, USP is used as a slow release acidifier excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₁₀O₆ MW: 178.14 g/mol Boiling Pt: 385 Melting Pt: 153 Storage Temperature: Ambient |
MDL Number:
MFCD00006647 CAS Number: 90-80-2 Merck Index: 13,04470 |
Specification Test Results
Assay | 99.0 - 101.0% |
Lead (Pb) | max. 0.001% |
Heavy Metals | max. 0.002% |
Reducing Substances | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...