Fuller's earth, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

F1093-2.5KG F1093-45KGBL F1093-500GM F1093-12KG
700000-346EA 249.02 USD
700000-346 700000-348 700000-350 700000-344
Fuller's earth, powder
Fuller's earth

Fuller's Earth, Powder is a palygorskite or bentonite clay material that has the ability to decolorize oil and other liquids. It is typically used as an absorbent for grease or oil.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00081617
CAS Number: 8031-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR