Ferric subsulfate solution USP

Supplier: SPECTRUM CHEMICAL MFG CORP

F1305-500CS6 F1305-200LTB F1305-500ML
95033-222CS 1607.7 USD
95033-222 700001-556 95033-224
Ferric subsulfate solution USP
Ferric subsulfate

Ferric Subsulfate Solution, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: ∼Fe₄(OH)₂(SO₄)₅
Density: 1.548
Storage Temperature: Ambient
CAS Number: 1310-45-8

Order Now

Specification Test Results

Assay (Fe) 20 - 22 G/100 ML
Nitrate (NO₃) To Pass Test
Ferrous Salts To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR