Ferric subsulfate, powder, purified
Supplier: SPECTRUM CHEMICAL MFG CORP
F1042-45KGBL
F1042-125GM
F1042-2.5KG
F1042-500GM
F1042-12KG
700001-548EA
4829.45
USD
700001-548
700001-542
700001-546
700001-550
700001-544
Ferric subsulfate, powder, purified
Ferric subsulfate
Ferric Subsulfate, Powder, Purified is a yellow rhombic crystalline salt that is water soluble. It is used in industrial waste processing as a coagulant and as a pigment in pickling baths for steel and aluminum.
Formula:
∼Fe₄(OH)₂(SO₄)₅ Storage Temperature: Ambient |
CAS Number:
1310-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...