Ferric pyrophosphate, Powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Iron(III) pyrophosphate, Iron (III) pyrophosphate

F1036-2.5KG F1036-500GM F1036-12KG
95033-218EA 1021.65 USD
95033-218 95033-220 700001-764
Ferric pyrophosphate, Powder FCC
Ferric pyrophosphate

Ferric Pyrophosphate, Powder, FCC is used as an iron fortification food supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016091
CAS Number: 10058-44-3

Order Now

Specification Test Results

Assay (Fe) 24.0-26.0 %
Arsenic (As) max.3 mg/kg
Lead (Pb) max. 4 mg/kg
Loss On Ignition max. 20.0 %
Mercury max. 3 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR