Fennel oil FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

FE135-100ML FE135-200LTB FE135-500ML
700010-626EA 194 USD
700010-626 700010-628 75811-206
Fennel oil FCC
Fennel oil

Fennel Oil, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Storage Temperature: Ambient
MDL Number: MFCD00146918
CAS Number: 8006-84-6

Order Now

Specification Test Results

Angular Rotation +12 to +24
Refractive Index @ 20°C 1.532 - 1.543
Solidification Point 3°C min.
Solubility in Alcohol To Pass Test
Specific Gravity @ 25°C 0.953 - 0.973


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR