Sunset yellow FCF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium 6-hydroxy-5-((4-sulphonatophenyl)diazenyl)naphthalene-2-sulphonate, Food yellow No. 5, FD and C Yellow No. 6
FD155-2.5KG
FD155-20KGBL
FD155-100GM
FD155-500GM
700010-078EA
819.5
USD
700010-078
700010-080
700010-076
700010-082
Sunset yellow FCF
Sunset yellow FCF
FD and C Yellow No. 6 | Non-graded FD and C Yellow No. 6 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₁₀N₂Na₂O₇S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00036437 CAS Number: 2783-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...