Ethyl cellulose, 10 mPa.s NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethylcellulose

ET110-125GM ET110-2.5KG ET110-500GM ET110-12KG ET110-45KGBL
95033-172EA 213.27 USD
95033-172 95033-174 95033-176 700012-994 700012-996
Ethyl cellulose, 10 mPa.s NF
Ethyl cellulose

Ethylcellulose, 10 mPa.s, NF. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now

Specification Test Results

Assay (Ethoxy Groups: -OC₂H₅; Dried Basis) 44.0 - 51.0 %
Capillary Viscosity 8.0 - 12.0 mPa.s
Acidity or Alkalinity To Pass Test
Loss On Drying max. 3.0 %
Residue On Ignition max. 0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR